Thermo Scientific Chemicals

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals

產品號碼: 212440250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals

產品號碼: 212440250
25 g, Glass bottle, Each
Quantity
Packaging
產品號碼: 212440250
價格 (TWD)

化學識別

CAS
69365-72-6
IUPAC Name
gallium(3+) trinitrate
Molecular Formula
GaN3O9
InChI Key
CHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Form)
Crystalline powder or crystals and/or chunks
Total trace metal impurities
=<0.0002 %
Appearance (Color)
White
Gallium (Ga)
18 to 22 %

說明

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text