Thermo Scientific Chemicals

Cloruro de difenilfosfinilo, 98 %, Thermo Scientific Chemicals

Número de catálogo: 212960100
10 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

Cloruro de difenilfosfinilo, 98 %, Thermo Scientific Chemicals

Número de catálogo: 212960100
10 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 212960100
Precio (USD)

Identificadores químicos

CAS
1499-21-4
IUPAC Name
diphenylphosphinoyl chloride
Molecular Formula
C12H10ClOP
InChI Key
QPQGTZMAQRXCJW-UHFFFAOYSA-N
SMILES
ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.6080 to 1.6120 (20°C, 589 nm)

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text