EZ-Link™ Sulfo-NHS-LC-Biotin
EZ-Link™ Sulfo-NHS-LC-Biotin
EZ-Link™ Sulfo-NHS-LC-Biotin
EZ-Link™ Sulfo-NHS-LC-Biotin
Thermo Scientific™
EZ-Link™ Sulfo-NHS-LC-Biotin
Thermo Scientific EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primaryRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21335100 mg
A3925710 x 1 mg
Catalog number 21335
Price (MXN)
-
Quantity:
100 mg
Request bulk or custom format
Thermo Scientific EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
Features of EZ-Link Sulfo-NHS-LC-Biotin:
• Protein labeling—biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
• Cell surface labeling—biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
• Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
• Soluble—charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
• Irreversible—forms permanent amide bonds; spacer arm cannot be cleaved
• Medium length—spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass
Sulfo-NHS-LC-Biotin is one of three very similar EZ-Link Reagents that are water-soluble, non-cleavable, and enable simple and efficient biotinylation of antibodies, proteins and any other primary amine-containing macromolecules in solution. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents. Differing only in their spacer arm lengths, the three Sulfo-NHS-ester reagents offer the possibility of optimizing labeling and detection experiments where steric hindrance of biotin binding is an important factor. Sulfo-NHS-LC-Biotin is offered in several package sizes and as complete protein labeling kits.
We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency, and performance for the intended research applications.
N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins (e.g., antibodies) typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide.
Varieties of biotin NHS-ester reagents differ in length, solubility, cell permeability and cleavability. Non-sulfonated NHS-biotins are cell permeable but must be dissolved in organic solvent such as DMSO or DMF. Sulfo-NHS biotins (and those with pegylated spacers) are directly water soluble but not membrane permeable. Varieties containing disulfide bonds can be cleaved using reducing agents, enabling the biotin group to be disconnected from the labeled protein.
Related Products
EZ-Link™ Sulfo-NHS-LC-Biotinylation Kit
EZ-Link™ Micro Sulfo-NHS-LC-Biotinylation Kit
Pierce™ Premium Grade Sulfo-NHS-LC-Biotin
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Impermeant
DescriptionEZ-Link Sulfo-NHS-LC-Biotin
Label TypeBiotin & Analogs
Product LineEZ-Link
Product TypeSulfo-NHS-LC-Biotin
Quantity100 mg
Reactive MoietyActive Ester, Succinimidyl Ester, Sulfo-NHS Ester
Chemical ReactivityAmine
Label or DyeBiotin
SolubilityDMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
SpacerMid-length
Unit SizeEach
Contents & Storage
Store desiccated at -20°C. Shipped at ambient temperature.
Frequently asked questions (FAQs)
Can I make a Sulfo-NHS-LC-Biotin stock solution to use later?
No. NHS esters hydrolyze very quickly. The container must be brought to room temperature (RT) prior to opening, then the amount of biotinylation reagent weighed, dissolved and used immediately.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can Sulfo-NHS-LC-Biotin be used in cell surface biotinylation?
Yes. Because this molecule dissolves readily in polar solutions and is itself charged by the sodium sulfoxide group on the succinimidyl ring, it cannot permeate the cell wall or membrane of an intact cell. As long as the cell is intact, only the primary amines on the surface will be biotinylated.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Why can't I perform biotinylation reactions in Tris buffer with Sulfo-NHS-LC-Biotin?
Tris is a primary amine, which will directly compete with protein primary amines for reaction with the NHS-ester activated biotin reagent. However, once a protein is labeled and desalted, Tris buffer can be used for storage or assay buffers, as appropriate for the protein and application.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What molar excess of Sulfo-NHS-LC-Biotin should I use to label antibodies and other proteins?
The molar excess to use depends on the concentration of the ligand to be biotinylated. Although the literature contains references that use between 2-50 molar excess of biotin, we recommend the following: 10-fold molar excess for 10 mg/mL solution of protein; 25-fold molar excess for 2 mg/mL solution of protein.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I dissolve the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257, 21335) in water for storage?
No, the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257) should not be dissolved in water for storage. We recommended storing EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format at -20 degrees C with desiccant as it is very moisture sensitive. Furthermore, to avoid moisture condensation onto the product, equilibrate vial to room temperature before opening.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Citations & References (15)
Citations & References
Abstract
Mechanisms of tethering and cargo transfer during epididymosome-sperm interactions.
Authors:Zhou W, Stanger SJ, Anderson AL, Bernstein IR, De Iuliis GN, McCluskey A, McLaughlin EA, Dun MD, Nixon B
Journal:BMC Biol
PubMed ID:30999907
'The mammalian epididymis is responsible for the provision of a highly specialized environment in which spermatozoa acquire functional maturity and are subsequently stored in preparation for ejaculation. Making important contributions to both processes are epididymosomes, small extracellular vesicles released from the epididymal soma via an apocrine secretory pathway. While considerable ... More
Molecular determinants in Frizzled, Reck, and Wnt7a for ligand-specific signaling in neurovascular development.
Authors:Cho C, Wang Y, Smallwood PM, Williams J, Nathans J
Journal:Elife
PubMed ID:31225798
'The molecular basis of Wnt-Frizzled specificity is a central question in developmental biology. Reck, a multi-domain and multi-functional glycosylphosphatidylinositol-anchored protein, specifically enhances beta-catenin signaling by Wnt7a and Wnt7b in cooperation with the 7-transmembrane protein Gpr124. Among amino acids that distinguish Wnt7a and Wnt7b from other Wnts, two clusters are essential ... More
Norrin-induced Frizzled4 endocytosis and endo-lysosomal trafficking control retinal angiogenesis and barrier function.
Authors:Zhang C, Lai MB, Khandan L, Lee LA, Chen Z, Junge HJ
Journal:Nat Commun
PubMed ID:28675177
Angiogenesis and blood-brain barrier formation are required for normal central nervous system (CNS) function. Both processes are controlled by Wnt or Norrin (NDP) ligands, Frizzled (FZD) receptors, and ß-catenin-dependent signalling in vascular endothelial cells. In the retina, FZD4 and the ligand NDP are critical mediators of signalling and are mutated ... More
USP48 restrains resection by site-specific cleavage of the BRCA1 ubiquitin mark from H2A.
Authors:Uckelmann M, Densham RM, Baas R, Winterwerp HHK, Fish A, Sixma TK, Morris JR
Journal:Nat Commun
PubMed ID:29335415
BRCA1-BARD1-catalyzed ubiquitination of histone H2A is an important regulator of the DNA damage response, priming chromatin for repair by homologous recombination. However, no specific deubiquitinating enzymes (DUBs) are known to antagonize this function. Here we identify ubiquitin specific protease-48 (USP48) as a H2A DUB, specific for the C-terminal BRCA1 ubiquitination ... More
The IgCAM CLMP regulates expression of Connexin43 and Connexin45 in intestinal and ureteral smooth muscle contraction in mice.
Authors:Langhorst H, Jüttner R, Groneberg D, Mohtashamdolatshahi A, Pelz L, Purfürst B, Schmidt-Ott KM, Friebe A, Rathjen FG
Journal:Dis Model Mech
PubMed ID:29361518
CAR-like membrane protein (CLMP), an immunoglobulin cell adhesion molecule (IgCAM), has been implicated in congenital short-bowel syndrome in humans, a condition with high mortality for which there is currently no cure. We therefore studied the function of CLMP in a ... More