Product Image
Thermo Scientific™
EZ-Link™ Micro Sulfo-NHS-LC-Biotinylation Kit
The Thermo Scientific EZ-Link Sulfo-NHS-LC-Biotinylation Kit contains reagents sufficient for 8 biotinylation reactions (e.g., 0.05–0.2 mg antibody per reaction). TheRead more
Have Questions?
Catalog NumberQuantity
219358 Reactions
Catalog number 21935
Price (USD)
548.00
Each
Quantity:
8 Reactions
Request bulk or custom format
Price (USD)
548.00
Each
The Thermo Scientific EZ-Link Sulfo-NHS-LC-Biotinylation Kit contains reagents sufficient for 8 biotinylation reactions (e.g., 0.05–0.2 mg antibody per reaction). The EZ-Link Sulfo-NHS-LC-Biotin reagent included in the kit is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins, and other molecules that have primary amines.
Features of EZ-Link Sulfo-NHS-LC-Biotin:
• Protein labeling—biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
• Cell surface labeling—biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
• Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
• Soluble—charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
• Irreversible—forms permanent amide bonds; spacer arm cannot be cleaved
• Medium length—spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass
Sulfo-NHS-LC-Biotin is one of three very similar EZ-Link Reagents that are water-soluble, non-cleavable, and enable simple and efficient biotinylation of antibodies, proteins and any other primary amine-containing macromolecules in solution. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents. Differing only in their spacer arm lengths, the three Sulfo-NHS-ester reagents offer the possibility of optimizing labeling and detection experiments where steric hindrance of biotin binding is an important factor. Sulfo-NHS-LC-Biotin is offered in several package sizes and as complete protein labeling kits.
We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency, and performance for the intended research applications.
N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins (e.g., antibodies) typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide.
Varieties of biotin NHS-ester reagents differ in length, solubility, cell permeability and cleavability. Non-sulfonated NHS-biotins are cell permeable but must be dissolved in organic solvent such as DMSO or DMF. Sulfo-NHS biotins (and those with pegylated spacers) are directly water soluble but not membrane permeable. Varieties containing disulfide bonds can be cleaved using reducing agents, enabling the biotin group to be disconnected from the labeled protein.
Related Products
EZ-Link™ Sulfo-NHS-LC-Biotin
EZ-Link™ Sulfo-NHS-LC-Biotinylation Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionEZ-Link Micro Sulfo-NHS-LC-Biotinylation Kit
Labeling Scale100 μg
Product LineEZ-Link
Product TypeMicro Sulfo-NHS-LC-Biotinylation Kit
Quantity8 Reactions
Labeling TargetAntibodies, Proteins
Label or DyeBiotin
SolubilityWater
Unit SizeEach
Contents & Storage
Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link Sulfo-NHS-LC-Biotin, 8 x 1 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns
Store biotin reagent at -20°C. Store remaining kit components at 4°C.
Frequently asked questions (FAQs)
In the EZ-Link Sulfo-NHS-LC-Biotinylation kit, what groups does Sulfo-NHS-LC-Biotin react with for biotinylation?
The NHS ester on this biotinylation reagent reacts with primary amines. In proteins and peptides, these occur at the amino terminus and the epsilon amine (side chain) of lysine residues.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Which EZ-Link Micro Biotinylation Kit is best for my application?
All four Micro Biotinylation Kits will label proteins for use in many applications. Consider the following guidelines for choosing among the kits:
• Choose the Sulfo-NHS-Biotinylation Kit (Cat. No. 21925) when it is important to minimize the molecular mass contributed by the label.
• Choose the Sulfo-NHS-SS-Biotinylation Kit (Cat. No. 21945) only when the downstream application require cleaving the biotin tag with a reducing agent.
• Choose the NHS-PEG4-Biotinylation Kit (Cat. No. 21955) to maximize protein solubility and prevent protein aggregation during storage.
• Choose the Sulfo-NHS-LC-Biotinylation Kit (Cat. No. 21935) for routine purposes when a feature of one of the other reagents is not required. The longer spacer arm offers the possibility of optimizing detection experiments where steric hindrance of biotin binding is an important factor.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How long does it take to biotinylate a protein using the EZ-Link Micro Biotinylation kits?
The entire procedure can be completed in less than 3 hours.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What types of proteins can be labeled with the EZ-Link Micro Biotinylation kits?
Any molecule containing a primary amine can be labeled. Any protein that has been labeled using the standard biotinylation kits, including antibodies and peptides, also can be labeled with the micro kits.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Are the biotinylation reagents in the EZ-Link Micro Biotinylation Kits cleavable?
Only the sulfo-NHS-SS-Biotin is cleavable because it has a disulfide bond in its spacer arm, enabling labeled proteins to be cleaved from the biotin group by treatment with reducing agents.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Citations & References (2)
Citations & References
Abstract
Antibody Microarray Immunoassay for Simultaneous Quantification of Multiple Mycotoxins in Corn Samples.
Authors:Zhang X, Wang Z, Fang Y, Sun R, Cao T, Paudyal N, Fang W, Song H
Journal:Toxins (Basel)
PubMed ID:30326616
We developed and tested a prototype of an antibody microarray immunoassay for simultaneous quantitative detection of four typical mycotoxins (aflatoxin B1, ochratoxin A, zearalenone, and fumonisin B1) in corn samples. The test kit consisted of a nitrocellulose membrane layered with immobilized monoclonal antibodies against mycotoxins. During the assay, the mycotoxin-protein ... More
Development of a Sensitive Chemiluminescence Immunoassay for the Quantification of Folic Acid in Human Serum.
Authors:Chen X, Zhou Q, Zhang T, Wang C, Yu Z, Ahamada H, Bai Z, Huang X
Journal:J Anal Methods Chem
PubMed ID:31240147
Folic acid (FA) is an important vitamin for human growth, especially for pregnant women. FA deficiency is associated with megaloblastic anemia, neural tube defects, cardiovascular diseases, irritability, diarrhea, and psychiatric disorders. Normally, FA molecules bind to folate-binding protein (FBP) in the serum as complex. Before quantify the FA concentration, a ... More