Methyl laurate, 98+%
Methyl laurate, 98+%
Methyl laurate, 98+%
Thermo Scientific Chemicals

Methyl laurate, 98+%

CAS: 111-82-0 | C13H26O2 | 214.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 215560100
Price (EUR)
31,20
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
31,20
Each
Chemical Identifiers
CAS111-82-0
IUPAC Namemethyl dodecanoate
Molecular FormulaC13H26O2
InChI KeyUQDUPQYQJKYHQI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4290 to 1.4340 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only