Lithium sulfate monohydrate, 99+%, pure
Lithium sulfate monohydrate, 99+%, pure
Lithium sulfate monohydrate, 99+%, pure
Lithium sulfate monohydrate, 99+%, pure
Thermo Scientific Chemicals

Lithium sulfate monohydrate, 99+%, pure

CAS: 10102-25-7 | H2Li2O5S | 127.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 218330010
Price (USD)
202.65
Special offer
Online exclusive
Ends: 28-Dec-2026
238.00
Save 35.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
202.65
Special offer
Online exclusive
Ends: 28-Dec-2026
238.00
Save 35.35
Each
Chemical Identifiers
CAS10102-25-7
IUPAC Namedilithium(1+) hydrate sulfate
Molecular FormulaH2Li2O5S
InChI KeyRKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES[Li+].[Li+].O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration after Ion exchange>=99.0 % (On dry substance)
Loss on ignition13.5 % to 15.0 % (600°C, 2 h)
Appearance (Color)White

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only