LC-SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxy-(6-amidocaproate))
Thermo Scientific™

LC-SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxy-(6-amidocaproate))

Thermo Scientific Pierce LC-SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of aRead more
Have Questions?
Catalog NumberQuantity
2236250 mg
Catalog number 22362
Price (USD)
231.00
Each
Quantity:
50 mg
Price (USD)
231.00
Each
Thermo Scientific Pierce LC-SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a long cyclohexane-stabilized spacer arm (16.2 angstroms).

Features of LC-SMCC:

• Reactive groups: NHS ester and maleimide
• Reactive toward: amino and sulfhydryl groups
• Long-chain variety of SMCC
• High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
• Non-cleavable
• Water-insoluble (dissolve first in DMF or DMSO)
• Cyclohexane bridge confers added stability to the maleimide group making LC-SMCC an ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C.

LC-SMCC is a non-cleavable and membrane permeable crosslinker. It contains an amine-reactive N-hydroxysuccinimide (NHS ester) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of LC-SMCC and are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm.

Applications:
• Ideal reagent for enzyme labeling of antibodies, both enzyme activity and antibody specificity can be preserved
• Create specific bioconjugates via one- or two-step crosslinking reactions
• Create sulfhydryl-reactive, maleimide-activated carrier proteins for coupling haptens

LC-SMCC is often used for the preparation of antibody-enzyme and hapten-carrier conjugates. In this type of conjugation, the NHS ester is reacted first with the antibody, excess crosslinking reagent removed and then the sulfhydryl-containing enzyme molecule is added. This two-step reaction scheme results in formation of specific antibody-enzyme conjugates.

LC-SMCC is a reagent of choice for creating stable maleimide-activated carrier proteins that will spontaneously react with sulfhydryls. In this application, the NHS ester of LC-SMCC is reacted with lysine residues on the carrier protein, converting them to reactive maleimides. These relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.

Specifications:
We manufacture LC-SMCC to the highest specifications to produce the most specific bioconjugates and maleimide-activated proteins, ensure the integrity of your data and to provide you with the highest degree of crosslinking consistency. Each lot of LC-SMCC is tested to meet the following minimum specifications.

• Identity: IR scan shows only peaks characteristic of the structure and functional groups of LC-SMCC crosslinker
• Purity: ≥ 90% by NMR

Product References:

Crosslinker Application Guide -- search for recent literature references for this product

Related Products
SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)
Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityYes
DescriptionLC-SMCC
FormSolid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)447.48
PEGylatedNo
Product LinePierce
Quantity50 mg
Reactive MoietyMaleimide, NHS Ester
Shipping ConditionAmbient
SolubilityDMF, DMSO
Spacer Arm Length16.2 Å
Water SolubleNo
Chemical ReactivityAmine-Sulfhydryl
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for LC-SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxy-(6-amidocaproate)))?

LC-SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxy-(6-amidocaproate)) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (3)

Citations & References
Abstract
BCL-2 selectively interacts with the BID-induced open conformer of BAK, inhibiting BAK auto-oligomerization.
Authors:Ruffolo SC, Shore GC
Journal:J Biol Chem
PubMed ID:12721291
'Caspase-8 cleaves BID to tBID, which targets mitochondria and induces oligomerization of BAX and BAK within the outer membrane, resulting in release of cytochrome c from the organelle. Here, we have initiated these steps in isolated mitochondria derived from control and BCL-2-overexpressing cells using synthetic BH3 peptides and subsequently analyzed ... More
Small molecule obatoclax (GX15-070) antagonizes MCL-1 and overcomes MCL-1-mediated resistance to apoptosis.
Authors:Nguyen M, Marcellus RC, Roulston A, Watson M, Serfass L, Murthy Madiraju SR, Goulet D, Viallet J, Bélec L, Billot X, Acoca S, Purisima E, Wiegmans A, Cluse L, Johnstone RW, Beauparlant P, Shore GC
Journal:Proc Natl Acad Sci U S A
PubMed ID:18040043
'Elevated expression of members of the BCL-2 pro-survival family of proteins can confer resistance to apoptosis in cancer cells. Small molecule obatoclax (GX15-070), which is predicted to occupy a hydrophobic pocket within the BH3 binding groove of BCL-2, antagonizes these members and induces apoptosis, dependent on BAX and BAK. Reconstitution ... More
Hybridization of an immobilized PNA probe with its complementary oligodeoxyribonucleotide on the surface of silica glass.
Authors:Masuko M
Journal:Nucleic Acids Res Suppl
PubMed ID:14510422
A 16mer sequence of peptide nucleic acid (PNA) was immobilized onto the surface of silica glass to supply solid supports for DNA chip and biosensor constructions. To the surface, 3-aminopropyltriethoxysilane (APTES), succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxy-(6-amidocaproate) (LC-SMCC), and cysteinyl-PNA were introduced in order. Hybridization of the immobilized PNA with its complementary oligodeoxyribonucleotide (ODN) should ... More