Ethanol, for spectroscopy, anhydrous, denat. with 5% 2-Propanol + 5% Methanol
Ethanol, for spectroscopy, anhydrous, denat. with 5% 2-Propanol + 5% Methanol
Ethanol, for spectroscopy, anhydrous, denat. with 5% 2-Propanol + 5% Methanol
Thermo Scientific Chemicals

Ethanol, for spectroscopy, anhydrous, denat. with 5% 2-Propanol + 5% Methanol

CAS: 64-17-5 | C2H6O | 46.069 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 224090025
Price (EUR)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<5 APHA
GC4.0 to 6.0 % (2-Propanol)
GC>=90.0 % (Ethanol)
UVat 205 nm A: =<1
Residue after evaporation=<0.0005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopy
For Manufacturing and Laboratory Use. Not for beverage use