Xylitol, 99+%
Xylitol, 99+%
Xylitol, 99+%
Thermo Scientific Chemicals

Xylitol, 99+%

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 225985000
Price (USD)
94.90
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
94.90
Each
Chemical Identifiers
CAS87-99-0
IUPAC Namepentane-1,2,3,4,5-pentol
Molecular FormulaC5H12O5
InChI KeyHEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point92°C to 96°C
Infrared spectrumConforms
Arsenic (As)=<3 ppm
GC=<2.0 % Other polyols
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only