This product ships via ground from US northeast.
L-Methionine sulfoximine, 98+%
L-Methionine sulfoximine, 98+%
L-Methionine sulfoximine, 98+%
Thermo Scientific Chemicals

L-Methionine sulfoximine, 98+%

CAS: 15985-39-4 | C5H12N2O3S | 180.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 227200050
Price (USD)
961.65
Online exclusive
1,068.00
Save 106.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
961.65
Online exclusive
1,068.00
Save 106.35
Each
Chemical Identifiers
CAS15985-39-4
IUPAC Name(2S)-2-azaniumyl-4-[imino(methyl)oxo-λ⁶-sulfanyl]butanoate
Molecular FormulaC5H12N2O3S
InChI KeySXTAYKAGBXMACB-DPVSGNNYSA-N
SMILESCS(=N)(=O)CC[C@H]([NH3+])C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO498.0 to 102.0 %
Appearance (Color)White to almost white
Specific optical rotation+12° to +15° (20°C, 589 nm) (c=1, H2O)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only