Pierce™ Dye Removal Columns
Thermo Scientific™

Pierce™ Dye Removal Columns

Thermo Scientific Pierce Dye Removal Columns effectively bind to unconjugated fluorescent dye molecules from protein solutions to rapidly purify fluorescentRead more
Have Questions?
Catalog NumberQuantity
2285810 columns, 5 mL resin
Catalog number 22858
Price (USD)
205.00
Each
Quantity:
10 columns, 5 mL resin
Price (USD)
205.00
Each
Thermo Scientific Pierce Dye Removal Columns effectively bind to unconjugated fluorescent dye molecules from protein solutions to rapidly purify fluorescent conjugated antibodies and other proteins after labeling reactions.

These Fluorescent Dye Removal Columns enable fast and efficient removal of non-reacted fluorescent dyes from protein labeling reactions. Removing excess dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. The dye removal resin in this kit is highly specialized to produce exceptional protein recoveries while effectively removing non-conjugated dye. Using the appropriate amount of resin and buffer conditions, almost any fluorescent dye can be removed with this kit.

Features of Dye Removal Columns:

• Fast—removes free, unconjugated dye from labeled protein solutions in less than a minute
• Specific—purification resin provides outstanding conjugate recovery (75 to 95%)
• Customizable—separate resin and microcentrifuge columns allow optimization for different dyes and concentrations of sample
• Sample-friendly—processing results in minimal sample dilution

This product has been used successfully to remove excess, unconjugated fluorescein, rhodamine and DyLight Dyes from antibodies following labeling reactions. Samples with dyes in the green spectrum, such as Dylight 549 and rhodamine, might require double processing because they require a high molar excesses for labeling. When using fluorescent dyes other than those described in the product instructions, the ratio of resin to protein/dye mixture will require optimization.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit, Microcentrifuge, Spin Column
Product TypeDye Removal Column
Quantity10 columns, 5 mL resin
TargetFluorescent Dye Removal
Product LinePierce
Unit SizeEach
Contents & Storage
Store purification resin at 4°C. Shipped at ambient temperature.

Frequently asked questions (FAQs)

Can the Dye Removal Columns be reused?

No. The resin is effective for one-time use only and cannot be regenerated.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is the Dye Removal resin solvent-compatible?

Yes, the resin is compatible with up to 10% organic solvent such as DMF.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is there sodium azide in the Dye Removal resin's storage solution?

Yes. It contains 0.02% sodium azide.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Which fluorescent dyes can I remove with the Dye Removal resin?

DyLight Dyes, rhodamine and fluorescein have been removed successfully using this resin. Samples with dyes in the green spectrum, such as Dylight 488 Dyes, might require double processing because they require a high molar excesses for labeling. The ratio of resin to protein/dye mixture is typically 1:1. When using fluorescent dyes other than those indicated, the ratio of resin to protein/dye mixture will require optimization.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How many samples can I process with the Dye Removal Columns?

The kit includes sufficient material to process 10 samples of 250 µl each for most dyes. The kit contains 5 mL of Dye Removal Resin, 10 Spin Columns and 20 Collection Tubes. The ideal labeled-protein concentration range is 1-2.5 mg/mL.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.