Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Thermo Scientific Chemicals

Benzo-18-crown-6, 97%

CAS: 14098-24-9 | C16H24O6 | 312.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 243980050
Price (EUR)
134,00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
134,00
Each
Add to cart
Chemical Identifiers
CAS14098-24-9
IUPAC Name2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Molecular FormulaC16H24O6
InChI KeyDSFHXKRFDFROER-UHFFFAOYSA-N
SMILESC1COCCOCCOC2=CC=CC=C2OCCOCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point40°C to 45°C
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only