2,4,6-Trimethylbenzoyl chloride, 98%
2,4,6-Trimethylbenzoyl chloride, 98%
2,4,6-Trimethylbenzoyl chloride, 98%
Thermo Scientific Chemicals

2,4,6-Trimethylbenzoyl chloride, 98%

CAS: 938-18-1 | C10H11ClO | 182.65 g/mol
数量:
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 244280100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS938-18-1
IUPAC Name2,4,6-trimethylbenzoyl chloride
Molecular FormulaC10H11ClO
InChI KeyUKRQMDIFLKHCRO-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(Cl)=O)C(C)=C1
さらに表示
GC>=97.5 %
Refractive index1.5280 to 1.5300 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only