2-(2-Ethoxyethoxy)ethyl acetate, 99%
2-(2-Ethoxyethoxy)ethyl acetate, 99%
2-(2-Ethoxyethoxy)ethyl acetate, 99%
Thermo Scientific Chemicals

2-(2-Ethoxyethoxy)ethyl acetate, 99%

CAS: 112-15-2 | C8H16O4 | 176.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 257420025
Price (USD)
125.65
Online exclusive
139.00
Save 13.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
125.65
Online exclusive
139.00
Save 13.35
Each
Chemical Identifiers
CAS112-15-2
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only