Methyl cellulose, viscosity 4000 cP (2% solution in water)
Methyl cellulose, viscosity 4000 cP (2% solution in water)
Methyl cellulose, viscosity 4000 cP (2% solution in water)
Thermo Scientific Chemicals

Methyl cellulose, viscosity 4000 cP (2% solution in water)

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 258110025
Price (USD)
452.00
Each
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
452.00
Each
Chemical Identifiers
CAS9004-67-5
IUPAC Name3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC20H38O11
InChI KeyYLGXILFCIXHCMC-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on drying=<5 %
Appearance (Color)White to yellowish-white to greyish-white
Infrared spectrumConforms
Viscosity3000 to 5600 cP (2 % in water at 20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only