사양
인증/적합성All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
설명96F well plate, Non-Treated, clear, with lid, Sterile (Pack of 10, Case of 160)
형식Standard
뚜껑With Lid
케이스당 수160/Cs.
팩당 수10 Pk.
포장160 Cases
제품 유형Microplate
수량10 Pk.
어레이8 x 12
바코드No Barcode
색상Clear
물질Polystyrene
웰 수96
제품라인Nunc
멸균Sterile
표면 처리Untreated
웰 형상Flat Bottom (F-well)
Unit SizeCase of 160