L(-)-Camphorsulfonic acid, 98%
L(-)-Camphorsulfonic acid, 98%
L(-)-Camphorsulfonic acid, 98%
Acros Organics

L(-)-Camphorsulfonic acid, 98%

CAS: 35963-20-3 | C10H16O4S | 232.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
26364025025 gGlass bottle
Catalog number 263640250
HSN 29147090
Price (INR)
7,476.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,476.00
Each
Chemical Identifiers
CAS35963-20-3
IUPAC Name[(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Molecular FormulaC10H16O4S
InChI KeyMIOPJNTWMNEORI-MHPPCMCBSA-N
SMILESCC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Specific optical rotation-21° to -23° (20°C, 589 nm) (c=20, H2O)
Appearance (Form)Crystalline powder
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only