Oleic acid, 97%
Oleic acid, 97%
Oleic acid, 97%
Thermo Scientific Chemicals

Oleic acid, 97%

CAS: 112-80-1 | C18H34O2 | 282.47 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 270291000
価格(JPY)
-
見積もりを依頼する
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS112-80-1
IUPAC Name(9E)-octadec-9-enoic acid
Molecular FormulaC18H34O2
InChI KeyZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILESCCCCCCCC\C=C\CCCCCCCC(O)=O
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4580 to 1.4610 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only