1-Naphthoyl chloride, 99%
1-Naphthoyl chloride, 99%
1-Naphthoyl chloride, 99%
Thermo Scientific Chemicals

1-Naphthoyl chloride, 99%

CAS: 879-18-5 | C11H7ClO | 190.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 270380100
Price (EUR)
47,00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,00
Each
Chemical Identifiers
CAS879-18-5
IUPAC Namenaphthalene-1-carbonyl chloride
Molecular FormulaC11H7ClO
InChI KeyNSNPSJGHTQIXDO-UHFFFAOYSA-N
SMILESClC(=O)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to beige to orange-brown
Infrared spectrumConforms
Refractive index1.6510 to 1.6540 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only