(+)-Camptothecin, 98%
(+)-Camptothecin, 98%
(+)-Camptothecin, 98%
Thermo Scientific Chemicals

(+)-Camptothecin, 98%

CAS: 7689-03-4 | C20H16N2O4 | 348.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number 276720010
Price (EUR)
972,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
972,00
Each
Chemical Identifiers
CAS7689-03-4
IUPAC Name(19S)-19-ethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular FormulaC20H16N2O4
InChI KeyVSJKWCGYPAHWDS-FQEVSTJZSA-N
SMILESCC[C@@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC=CC1=C3)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<1.0 %
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only