(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 95+%
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 95+%
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 95+%
Thermo Scientific Chemicals

(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 95+%

CAS: 72748-99-3 | C6H14N2O | 130.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2771400101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 277140010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS72748-99-3
IUPAC Name(2R)-1-azaniumyl-2-(methoxymethyl)pyrrolidin-1-ium
Molecular FormulaC6H16N2O
InChI KeyCJGJJXCQCKRPHC-ZCFIWIBFSA-N
SMILESCOC[C@H]1CCC[NH+]1[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4635 to 1.4655 (20°C, 589 nm)
GC>=95.0 %
Specific optical rotation+76° to +84° (20°C, 589 nm) (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only