Polysorbate 60
Polysorbate 60
Polysorbate 60
Thermo Scientific Chemicals

Polysorbate 60

CAS: 9005-67-8 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | 522.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 278622500
Price (USD)
40.65
Online Exclusive
44.50
Save 3.85 (9%)
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
44.50
Save 3.85 (9%)
Each
Chemical Identifiers
CAS9005-67-8
Molecular Formula(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
InChI KeyDEKFZWMENCCOFU-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD01780110
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to light brown
Infrared spectrumConforms
Hydroxyl value81 to 96 mg KOH/g
Dioxane=<5 ppm
Acid value=<2 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only