This product ships via ground from US northeast.
Polysorbate 80
Polysorbate 80
Polysorbate 80
Thermo Scientific Chemicals

Polysorbate 80

Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Catalog number 278630100
Price (USD)
638.65
Online exclusive
709.00
Save 70.35
Each
Quantity:
10 L
Request bulk or custom format
Price (USD)
638.65
Online exclusive
709.00
Save 70.35
Each
Chemical Identifiers
CAS9005-65-6
Molecular Formula(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
InChI KeyHDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00082107
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4700 to 1.4760 (20°C, 589 nm)
Saponification value45 to 55 mg KOH/g
Hydroxyl value65 to 80 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Indoors +20°C