Search
| Katalognummer | Menge |
|---|---|
| 2943511050 | 50 ml |
| 2943511005 | 5 ml |
| 2943511010 | 10 ml |
CaptureSelect Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.
Features of this affinity matrix include:
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics: