Samarium, 99.9%, (trace metal basis), 20 to 40 mesh
Samarium, 99.9%, (trace metal basis), 20 to 40 mesh
Samarium, 99.9%, (trace metal basis), 20 to 40 mesh
Thermo Scientific Chemicals

Samarium, 99.9%, (trace metal basis), 20 to 40 mesh

CAS: 7440-19-9 | Sm | 150.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Catalog number 294780100
Price (USD)
199.65
Online Exclusive
222.00
Save 22.35 (10%)
Each
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
199.65
Online Exclusive
222.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS7440-19-9
IUPAC Namesamarium
Molecular FormulaSm
InChI KeyKZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES[Sm]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver grey
Total trace metal impurities=<0.1 %
Appearance (Form)Powder or filings
Particle size20 to 40 mesh
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only