3-Buten-1-ol, 98+%
3-Buten-1-ol, 98+%
3-Buten-1-ol, 98+%
Thermo Scientific Chemicals

3-Buten-1-ol, 98+%

CAS: 627-27-0 | C4H8O | 72.11 g/mol
数量:
25 mL
100 mL
500 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 295755000
価格(JPY)
-
見積もりを依頼する
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS627-27-0
IUPAC Namebut-3-en-1-ol
Molecular FormulaC4H8O
InChI KeyZSPTYLOMNJNZNG-UHFFFAOYSA-N
SMILESOCCC=C
さらに表示
Appearance (Form)Liquid
GC>=98.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only