1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
Thermo Scientific Chemicals

1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%

CAS: 41100-52-1 | C12H22ClN | 215.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 298080010
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.70
Save 13.05
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.70
Save 13.05
Each
Chemical Identifiers
CAS41100-52-1
IUPAC Namehydrogen 3,5-dimethyladamantan-1-amine chloride
Molecular FormulaC12H22ClN
InChI KeyLDDHMLJTFXJGPI-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO494 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only