4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
Thermo Scientific Chemicals

4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent

CAS: 5808-22-0 | C10H6O8S2 | 318.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 300600100
Price (USD)
69.65
Special offer
Online exclusive
Ends: 28-Dec-2026
81.50
Save 11.85
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 28-Dec-2026
81.50
Save 11.85
Each
Chemical Identifiers
CAS5808-22-0
IUPAC Name2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate
Molecular FormulaC10H10Na2O10S2
InChI KeyQUEAKWJKJBFNEG-UHFFFAOYSA-L
SMILESO.O.OC1=CC(=CC2=CC(=CC(O)=C12)S(=O)(=O)O[Na])S(=O)(=O)O[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Sensitivity(to nitrate) Passes test
Sensitivity(to formaldehyde) Passes test
Clarity of solutionPasses test
Appearance (Color)Off-white to beige to grey to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only