2-Chloroethylphosphonic acid, 90%, Tech.
2-Chloroethylphosphonic acid, 90%, Tech.
2-Chloroethylphosphonic acid, 90%, Tech.
Thermo Scientific Chemicals

2-Chloroethylphosphonic acid, 90%, Tech.

CAS: 16672-87-0 | C2H6ClO3P | 144.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 301630050
Price (USD)
621.65
Online exclusive
690.00
Save 68.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
621.65
Online exclusive
690.00
Save 68.35
Each
Chemical Identifiers
CAS16672-87-0
IUPAC Name(2-chloroethyl)phosphonic acid
Molecular FormulaC2H6ClO3P
InChI KeyUDPGUMQDCGORJQ-UHFFFAOYSA-N
SMILESOP(O)(=O)CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point65°C to 75°C
Appearance (Color)White to beige
Titration with NaOH>=88 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only