L-Lysine, 98%
L-Lysine, 98%
L-Lysine, 98%
Thermo Scientific Chemicals

L-Lysine, 98%

CAS: 56-87-1 | C6H14N2O2 | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number 303345000
Price (USD)
486.65
Special offer
Online exclusive
Ends: 28-Dec-2026
572.00
Save 85.35
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
486.65
Special offer
Online exclusive
Ends: 28-Dec-2026
572.00
Save 85.35
Each
Chemical Identifiers
CAS56-87-1
IUPAC Name(2S)-2,6-diaminohexanoic acid
Molecular FormulaC6H14N2O2
InChI KeyKDXKERNSBIXSRK-MLHKIVSYNA-N
SMILESNCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 % (on dry substance)
Specific optical rotation+23° to +27° (20°C, 589 nm) (c=2, 1N HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only