(S)-(+)-3-Hydroxytetrahydrofuran, 99%
(S)-(+)-3-Hydroxytetrahydrofuran, 99%
(S)-(+)-3-Hydroxytetrahydrofuran, 99%
Thermo Scientific Chemicals

(S)-(+)-3-Hydroxytetrahydrofuran, 99%

CAS: 86087-23-2 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 304430250
Price (USD)
186.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
186.00
Each
Chemical Identifiers
CAS86087-23-2
IUPAC Nameoxolan-3-ol
Molecular FormulaC4H8O2
InChI KeyXDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILESOC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Specific optical rotation+15° to +20° (20°C, 589 nm) (c=2, MeOH)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only