L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
Thermo Scientific Chemicals

L-Alanine methyl ester hydrochloride, 99%

CAS: 2491-20-5 | C4H10ClNO2 | 139.58 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 308360250
価格(JPY)
-
見積もりを依頼する
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2491-20-5
IUPAC Namemethyl (2S)-2-aminopropanoate hydrochloride
Molecular FormulaC4H10ClNO2
InChI KeyIYUKFAFDFHZKPI-DOIAFFBONA-N
SMILESCl.COC(=O)[C@H](C)N
さらに表示
Appearance (Color)White to off-white
Titration Argentometric>=98.5 %
Specific optical rotation+6° to +8° (24°C, 589 nm) (c=1.6, CH3OH)
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only