NeutrAvidin Protein, 1000 mg - FAQs
View additional product information for NeutrAvidin Protein - FAQs (31000, 31055, 31050)
1 product FAQs found
What is the shelf life of NeutrAvidin Protein?
NeutrAvidin Protein is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale for more details.