Methyl-beta-D-galactopyranoside, 98%
Methyl-beta-D-galactopyranoside, 98%
Methyl-beta-D-galactopyranoside, 98%
Thermo Scientific Chemicals

Methyl-beta-D-galactopyranoside, 98%

CAS: 1824-94-8 | C7H14O6 | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 314980050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1824-94-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
HPLC>=97.5 %
Infrared spectrumConforms
Water=<1 % (K.F.)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only