Cadmium acetate dihydrate, 98%, for analysis
Cadmium acetate dihydrate, 98%, for analysis
Cadmium acetate dihydrate, 98%, for analysis
Thermo Scientific Chemicals

Cadmium acetate dihydrate, 98%, for analysis

CAS: 5743-04-4 | C4H6CdO4·2H2O | 266.52 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 317135000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS5743-04-4
IUPAC Namecadmium(2+) diacetate dihydrate
Molecular FormulaC4H10CdO6
InChI KeyAUIZLSZEDUYGDE-UHFFFAOYSA-L
SMILESO.O.[Cd++].CC([O-])=O.CC([O-])=O
檢視更多
規格Specification Sheet規格表
Nitrate (NO3)=<30 ppm
Iron (Fe)=<10 ppm
Appearance (Color)White
Titration Complexometric>=97.5 %
Subst not pptd by H2S=<0.3 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only