Cyclobutyl bromide, 97%, pure
Cyclobutyl bromide, 97%, pure
Cyclobutyl bromide, 97%, pure
Thermo Scientific Chemicals

Cyclobutyl bromide, 97%, pure

CAS: 4399-47-7 | C4H7Br | 135.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 320500010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4399-47-7
IUPAC Namebromocyclobutane
Molecular FormulaC4H7Br
InChI KeyKXVUSQIDCZRUKF-UHFFFAOYSA-N
SMILESBrC1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4780 to 1.4810 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only