chlorodiisopropylphosphine, 96%
chlorodiisopropylphosphine, 96%
chlorodiisopropylphosphine, 96%
Thermo Scientific Chemicals

chlorodiisopropylphosphine, 96%

CAS: 40244-90-4 | C6H14ClP | 152.61 g/mol
Have Questions?
Catalog NumberQuantityPackaging
32155025025 gGlass bottle
Catalog number 321550250
Price (USD)
218.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
218.00
Each
Chemical Identifiers
CAS40244-90-4
IUPAC Namechlorobis(propan-2-yl)phosphane
Molecular FormulaC6H14ClP
InChI KeyJZPDBTOWHLZQFC-UHFFFAOYSA-N
SMILESCC(C)P(Cl)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only