Sodium tert-pentoxide, 95%, pure
Sodium tert-pentoxide, 95%, pure
Sodium tert-pentoxide, 95%, pure
Thermo Scientific Chemicals

Sodium tert-pentoxide, 95%, pure

CAS: 14593-46-5 | C5H11NaO | 110.13 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 327555000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14593-46-5
IUPAC Namesodium 2,2-dimethylpropan-1-olate
Molecular FormulaC5H11NaO
InChI KeyYQMLGVNRTAQUFQ-UHFFFAOYSA-N
SMILES[Na+].CC(C)(C)C[O-]
さらに表示
Titration with HClO4>=94 %
Appearance (Form)Powder
Appearance (Color)Cream to tan
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only