4-Amino-2-butanol, 98%
4-Amino-2-butanol, 98%
4-Amino-2-butanol, 98%
Thermo Scientific Chemicals

4-Amino-2-butanol, 98%

CAS: 39884-48-5 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 330230010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS39884-48-5
IUPAC Name(3R)-3-hydroxybutan-1-aminium
Molecular FormulaC4H12NO
InChI KeyNAXUFNXWXFZVSI-SCSAIBSYSA-O
SMILESC[C@@H](O)CC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4518 to 1.4538 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only