1-Naphthyl butyrate, 98%
1-Naphthyl butyrate, 98%
1-Naphthyl butyrate, 98%
Thermo Scientific Chemicals

1-Naphthyl butyrate, 98%

CAS: 3121-70-8 | C14H14O2 | 214.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 334950010
Price (USD)
69.65
Special offer
Online exclusive
Ends: 28-Dec-2026
82.00
Save 12.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 28-Dec-2026
82.00
Save 12.35
Each
Chemical Identifiers
CAS3121-70-8
IUPAC Namenaphthalen-1-yl butanoate
Molecular FormulaC14H14O2
InChI KeyXIVJNRXPRQKFRZ-UHFFFAOYSA-N
SMILESCCCC(=O)OC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only