4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%
4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%
4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%
Thermo Scientific Chemicals

4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%

CAS: 2465-59-0 | C5H4N4O2 | 152.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 339940010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2465-59-0
IUPAC Name1H,2H,4H,5H,6H-pyrazolo[3,4-d]pyrimidine-4,6-dione
Molecular FormulaC5H4N4O2
InChI KeyHXNFUBHNUDHIGC-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CNNC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Light yellow
Appearance (Form)Powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only