1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
Thermo Scientific Chemicals

1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized

CAS: 27905-45-9 | C13H7F17O2 | 518.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3486100505 gGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 348610050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS27905-45-9
IUPAC Name3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate
Molecular FormulaC13H7F17O2
InChI KeyQUKRIOLKOHUUBM-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
StabilizerApprox. 225 ppm MEHQ
Stabilizeror approx. 15 ppm TBC and 70 ppm BHT
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only