1-Aminobenzotriazole, 98%
1-Aminobenzotriazole, 98%
1-Aminobenzotriazole, 98%
Thermo Scientific Chemicals

1-Aminobenzotriazole, 98%

CAS: 1614-12-6 | C6H6N4 | 134.14 g/mol
数量:
100 mg
250 mg
パッケージ:
Glass bottle
製品番号(カタログ番号) 348642500
価格(JPY)
-
数量:
250 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1614-12-6
IUPAC Name1H-1,2,3-benzotriazol-1-amine
Molecular FormulaC6H6N4
InChI KeyJCXKHYLLVKZPKE-UHFFFAOYSA-N
SMILESNN1N=NC2=CC=CC=C12
さらに表示
Appearance (Color)Yellow to beige to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Melting point81°C to 85°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only