Celite™ 545
Thermo Scientific Chemicals

Celite™ 545

CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Fiber Drum
Catalog number 349670100
Price (USD)
266.65
Special offer
Online exclusive
Ends: 28-Dec-2026
313.00
Save 46.35
Each
Quantity:
10 kg
Packaging:
Fiber Drum
Request bulk or custom format
Price (USD)
266.65
Special offer
Online exclusive
Ends: 28-Dec-2026
313.00
Save 46.35
Each
Chemical Identifiers
CAS68855-54-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
AppearanceMay contain small black particles
pH9.0 to 11.0 (10 % slurry)
Wet density255 to 478 g/l
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only