3-Bromo-1-propanol, 97%
3-Bromo-1-propanol, 97%
3-Bromo-1-propanol, 97%
Thermo Scientific Chemicals

3-Bromo-1-propanol, 97%

CAS: 627-18-9 | C3H7BrO | 138.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 349700250
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Chemical Identifiers
CAS627-18-9
IUPAC Name3-bromopropan-1-ol
Molecular FormulaC3H7BrO
InChI KeyRQFUZUMFPRMVDX-UHFFFAOYSA-N
SMILESOCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Refractive index1.4860 to 1.4900 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only