Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CD3 Antibodies

          Invitrogen

          CD3e Monoclonal Antibody (145-2C11), PE-Cyanine5.5, eBioscience™

          98 Published Figures
          140 References
          View all (604) CD3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD3e Monoclonal Antibody (145-2C11), PE-Cyanine5.5, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (98)
          CD3e Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD3e Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD3e Antibody (35-0031-80) in Flow

          Staining of BALB/c splenocytes with Anti-Human/Mouse CD45R (B220) FITC (Product # 11-0452-82) and 0.125 µg of Armenian Hamster IgG Isotype Control PE-Cyanine5-5 (Product # 35-4888-80) (left) or 0.125 µg of Anti-Mouse CD3e PE-Cyanine5-5 (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          35-0031-80

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 137 publications 137 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), PE-Cyanine5.5, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          145-2C11

          Conjugate

          PE-Cyanine5.5 PE-Cyanine5.5 PE-Cyanine5.5
          • Unconjugated
          • Alexa Fluor 488 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 610-70S
          • NovaFluor Blue 660-120S
          • NovaFluor Red 685
          • NovaFluor Red 700
          • NovaFluor Red 710
          • NovaFluor Yellow 610
          • NovaFluor Yellow 660
          • NovaFluor Yellow 690
          • NovaFluor Yellow 700
          • NovaFluor Yellow 730
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          564/692 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11218085

          Product Specific Information

          Description: The 145-2C11 monoclonal antibody reacts with mouse CD3e, a 20 kDa subunit of the TCR complex. Along with the other CD3 subunits, gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 145-2C11 to TCR initiates the intracellular biochemical pathway resulting in cellular activation, proliferation, and apoptosis depending on specific conditions utilized. 145-2C11 is commonly used as a phenotypic marker for mouse T cells.

          Applications Reported: This 145-2C11 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 145-2C11 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 488-561 nm; Emission: 695 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Target Information

          The CD3 complex, composed of gamma, delta, epsilon, and zeta subunits, is essential for the assembly, trafficking, and surface expression of the T cell receptor (TCR) complex. These subunits are structurally related members of the immunoglobulin superfamily and are encoded by closely linked genes on human chromosome 11. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, but not on B or NK cells. The CD3 subunits play a crucial role in transducing antigen-recognition signals into the cytoplasm of T cells. The cytoplasmic tails of CD3 subunits contain a double tyrosine-based motif that associates with cytoplasmic signal transduction molecules, mediating T cell activation through the TCR. Crosslinking of the TCR initiates intracellular biochemical pathways that result in cellular activation, proliferation, and potentially growth arrest and cell survival. CD3 is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes, and Purkinje cells in the cerebellum. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Defects in the CD3 gene are associated with CD3 immunodeficiency, highlighting its importance in immune function and regulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD3 antigen, zeta polypeptide; CD3 TCR complex; CD3-zeta/eta; FLJ18683; Leu-4; T cell antigen receptor complex epsilon subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 eta chain; T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 zeta chain; T3-epsilon protein; T3/TCR complex; unnamed protein product

          View more View less

          Gene Aliases: 4930549J05Rik; A430104F18Rik; CD3; Cd3-eta; Cd3-zeta; CD3epsilon; Cd3h; Cd3z; Cd3zeta; Ctg-3; Ctg3; T3d; T3e; T3g; T3z; Tcrk; Tcrz

          View more View less

          UniProt ID: (Mouse) P24161, (Mouse) P04235, (Mouse) P22646, (Mouse) P11942

          View more View less

          Entrez Gene ID: (Mouse) 12503, (Mouse) 12500, (Mouse) 12501, (Mouse) 12502

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding protein binding, bridging MHC class II receptor activity identical protein binding protein homodimerization activity protein heterodimerization activity protein tyrosine kinase binding SH3 domain binding
          Process(es)
          positive regulation of cell-matrix adhesion positive regulation of T cell anergy cell surface receptor signaling pathway response to nutrient regulation of signal transduction positive regulation of gene expression negative regulation of gene expression dendrite development cerebellum development T cell costimulation positive regulation of interferon-gamma production positive regulation of interleukin-2 production positive regulation of interleukin-4 production positive regulation of cell-cell adhesion mediated by integrin positive regulation of T cell proliferation positive regulation of MAPK cascade positive thymic T cell selection negative thymic T cell selection negative regulation of smoothened signaling pathway positive regulation of alpha-beta T cell proliferation lymphocyte activation positive regulation of peptidyl-tyrosine phosphorylation positive regulation of calcium-mediated signaling T cell receptor signaling pathway positive regulation of T cell activation apoptotic signaling pathway T cell activation protein-containing complex assembly establishment or maintenance of cell polarity protein transport regulation of lymphocyte apoptotic process Fc-gamma receptor signaling pathway protein oligomerization positive regulation of protein localization to cell surface
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-v2vhp:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0