3-Bromophenethyl alcohol, 97%
3-Bromophenethyl alcohol, 97%
3-Bromophenethyl alcohol, 97%
Thermo Scientific Chemicals

3-Bromophenethyl alcohol, 97%

CAS: 28229-69-8 | C8H9BrO | 201.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 350560010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS28229-69-8
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5730 to 1.5750 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only