4-Ethyl-2-methoxyphenol, 98%
4-Ethyl-2-methoxyphenol, 98%
4-Ethyl-2-methoxyphenol, 98%
Thermo Scientific Chemicals

4-Ethyl-2-methoxyphenol, 98%

CAS: 2785-89-9 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 351960250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2785-89-9
IUPAC Name4-ethyl-2-methoxyphenol
Molecular FormulaC9H12O2
InChI KeyCHWNEIVBYREQRF-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.5250 to 1.5300 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only