Thermo Scientific Chemicals

n-Heneicosane, 98%, Thermo Scientific Chemicals

Catalog number: 352770100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

n-Heneicosane, 98%, Thermo Scientific Chemicals

Catalog number: 352770100
10 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
629-94-7
IUPAC Name
henicosane
Molecular Formula
C21H44
InChI Key
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCC
Appearance (Color)
White
Appearance (Form)
Crystalline chunks or crystals or flakes
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
39°C to 43°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text