N-Vinyl-2-pyrrolidone, 99%, stabilized with Kerobit(R), Thermo Scientific Chemicals
N-Vinyl-2-pyrrolidone, 99%, stabilized with Kerobit(R), Thermo Scientific Chemicals
N-Vinyl-2-pyrrolidone, 99%, stabilized with Kerobit(R), Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Vinyl-2-pyrrolidone, 99%, stabilized with Kerobit(R), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
3547300252.5 LGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 354730025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS88-12-0
IUPAC Name1-ethenylpyrrolidin-2-one
Molecular FormulaC6H9NO
InChI KeyWHNWPMSKXPGLAX-UHFFFAOYSA-N
SMILESC=CN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer5 to 15 ppm Kerobit(N,N'-Di-sec-butyl-p-phenylenediamine)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5110 to 1.5140 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only